Assay Details
| Gene Symbol: lon1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112433.1 | NP_001105903.1 | 1100 | 98 | ||
| GenBank mRNA | AY109454.1 | - | 1100 | 98 | ||
| U85494.1 | - | 1100 | 98 | |||
More Information
Set Membership: |
|

lon1
Genomic Map