Assay Details
| Gene Symbol: LOC100191856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137280.1 | NP_001130752.1 | 305 | 96 | ||
| GenBank mRNA | BT034089.1 | - | 294 | 96 | ||
| BT040833.1 | - | 222 | 96 | |||
| EU951879.1 | - | 305 | 96 | |||
More Information
Set Membership: |
|

LOC100191856
Genomic Map