Assay Details
Gene Symbol: LOC100191936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137360.1 | NP_001130832.1 | 383 | 99 | ||
GenBank mRNA | BT034194.1 | - | 383 | 99 | ||
BT067210.1 | - | 415 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
