Assay Details
Gene Symbol: LOC100192597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137812.1 | NP_001131284.1 | 291 | 70 | ||
GenBank mRNA | BT034619.1 | - | 274 | 70 | ||
EU958368.1 | - | 291 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
