Assay Details
Gene Symbol: LOC100193477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001138592.1 | NP_001132064.1 | 1055 | 99 | ||
GenBank mRNA | BT035745.1 | - | 885 | 99 | ||
EU961370.1 | - | 1055 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
