Assay Details
Gene Symbol: LOC100193569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001138678.1 | NP_001132150.1 | 1025 | 85 | ||
GenBank mRNA | BT035868.1 | - | 1025 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
