Assay Details
Gene Symbol: LOC100193689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001138781.1 | NP_001132253.1 | 474 | 103 | ||
GenBank mRNA | BT036021.1 | - | 474 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
