Assay Details
Gene Symbol: LOC100193998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139063.1 | NP_001132535.1 | 1545 | 61 | ||
GenBank mRNA | BT036409.1 | - | 1545 | 61 |
Gene Symbol: LOC100381294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174189.1 | NP_001167660.1 | 1331 | 61 | ||
GenBank mRNA | EU953392.1 | - | 1331 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
