Assay Details
Gene Symbol: LOC100194168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139218.1 | NP_001132690.1 | 218 | 96 | ||
GenBank mRNA | BT036630.1 | - | 172 | 96 | ||
BT037248.1 | - | 167 | 96 | |||
EU963519.1 | - | 218 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT037248.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
