Assay Details
Gene Symbol: LOC100194380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139414.1 | NP_001132886.1 | 1245 | 77 | ||
GenBank mRNA | BT036911.1 | - | 1245 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
