Assay Details
Gene Symbol: LOC100194325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139363.1 | NP_001132835.1 | 1289 | 64 | ||
GenBank mRNA | BT036842.1 | - | 1289 | 64 | ||
EU961417.1 | - | 1201 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
