Assay Details
Gene Symbol: LOC100194325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139363.1 | NP_001132835.1 | 366 | 79 | ||
GenBank mRNA | BT036842.1 | - | 366 | 79 | ||
BT041898.1 | - | 250 | 79 | |||
EU961417.1 | - | 278 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
