Assay Details
Gene Symbol: LOC100216611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143024.1 | NP_001136496.1 | 621 | 88 | ||
GenBank mRNA | BT037042.1 | - | 621 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
