Assay Details
Gene Symbol: LOC100216611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143024.1 | NP_001136496.1 | 825 | 73 | ||
GenBank mRNA | BT037042.1 | - | 825 | 73 | ||
EU676035.1 | - | 648 | 73 |
Gene Symbol: LOC100381806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174602.1 | NP_001168073.1 | 824 | 73 | ||
GenBank mRNA | BT062316.1 | - | 477 | 73 | ||
BT084109.1 | - | 824 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
