Assay Details
Gene Symbol: LOC100217142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143508.1 | NP_001136980.1 | 383 | 79 | ||
GenBank mRNA | BT038003.1 | - | 383 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
