Assay Details
Gene Symbol: LOC100217256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143611.1 | NP_001137083.1 | 541 | 73 | ||
GenBank mRNA | BT038218.1 | - | 541 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
