Assay Details
Gene Symbol: LOC100283480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156380.1 | NP_001149852.1 | 364 | 87 | ||
GenBank mRNA | EU964886.1 | - | 364 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
