Assay Details
| Gene Symbol: vdac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111479.1 | NP_001104949.1 | 208 | 102 | ||
| GenBank mRNA | AF178952.1 | - | 208 | 102 | ||
| BT040062.1 | - | 194 | 102 | |||
| EU962130.1 | - | 321 | 102 | |||
More Information
Set Membership: |
|

vdac2
Genomic Map