Assay Details
Gene Symbol: tbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111849.1 | NP_001105319.1 | 775 | 103 | ||
GenBank mRNA | AY109414.1 | - | 103 | |||
BT087853.1 | - | 834 | 103 | |||
EU951919.1 | - | 860 | 103 | |||
L13302.1 | - | 775 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
