Assay Details
Gene Symbol: LOC100194234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139275.1 | NP_001132747.1 | 551 | 79 | ||
GenBank mRNA | BT036725.1 | - | 551 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
