Assay Details
| Gene Symbol: rps6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112164.1 | NP_001105634.1 | 310 | 103 | ||
| GenBank mRNA | AF295599.1 | - | 310 | 103 | ||
| EU969025.1 | - | 334 | 103 | |||
More Information
Set Membership: |
|

rps6-2
Genomic Map