Assay Details
Gene Symbol: rab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112763.1 | NP_001106234.1 | 652 | 84 | ||
GenBank mRNA | BT068076.1 | - | 599 | 84 | ||
EU971297.1 | - | 684 | 84 | |||
U22433.1 | - | 652 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
