Assay Details
| Gene Symbol: LOC100281141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154060.1 | NP_001147532.1 | 970 | 98 | ||
| GenBank mRNA | BT041352.1 | - | 937 | 98 | ||
| EU955724.1 | - | 970 | 98 | |||
More Information
Set Membership: |
|

LOC100281141
Genomic Map