Assay Details
| Gene Symbol: nip2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111550.1 | NP_001105020.1 | 355 | 80 | ||
| GenBank mRNA | AF326485.1 | - | 355 | 80 | ||
| BT063648.1 | - | 351 | 80 | |||
| EU956287.1 | - | 371 | 80 | |||
More Information
Set Membership: |
|

nip2b
Genomic Map