Assay Details
Gene Symbol: pip2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111554.1 | NP_001105024.1 | 950 | 103 | ||
GenBank mRNA | AF326491.1 | - | 950 | 103 | ||
AY243801.1 | - | 879 | 103 | |||
BT033960.1 | - | 956 | 103 | |||
BT039587.1 | - | 949 | 103 | |||
BT041139.1 | - | 953 | 103 | |||
EU959656.1 | - | 1339 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
