Assay Details
| Gene Symbol: GRP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112096.1 | NP_001105566.1 | 245 | 103 | ||
| GenBank mRNA | AY095207.1 | - | 245 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRP5| Gene Symbol: GRP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112096.1 | NP_001105566.1 | 245 | 103 | ||
| GenBank mRNA | AY095207.1 | - | 245 | 103 | ||
Set Membership: |
|