Assay Details
Gene Symbol: pip2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111556.1 | NP_001105026.1 | 1053 | 103 | ||
GenBank mRNA | AF326494.1 | - | 1053 | 103 | ||
BT063385.1 | - | 1048 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
