Assay Details
Gene Symbol: cl5295_1(174) | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112558.1 | NP_001106028.1 | 748 | 111 | ||
GenBank mRNA | BT042513.1 | - | 849 | 111 | ||
BT087437.1 | - | 771 | 111 | |||
EF515849.1 | - | 748 | 111 | |||
EU960601.1 | - | 822 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
