Assay Details
Gene Symbol: LOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112505.1 | NP_001105975.1 | 2950 | 97 | ||
GenBank mRNA | BT063599.1 | - | 2880 | 97 | ||
DQ335763.1 | - | 2950 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
