Assay Details
| Gene Symbol: tap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138403.1 | NP_001131875.1 | 1043 | 101 | ||
| GenBank mRNA | BT035476.1 | - | 1043 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tap1| Gene Symbol: tap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138403.1 | NP_001131875.1 | 1043 | 101 | ||
| GenBank mRNA | BT035476.1 | - | 1043 | 101 | ||
Set Membership: |
|