Assay Details
| Gene Symbol: NIP2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112167.1 | NP_001105637.1 | 944 | 112 | ||
| GenBank mRNA | AF326484.1 | - | 944 | 112 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NIP2-1| Gene Symbol: NIP2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112167.1 | NP_001105637.1 | 944 | 112 | ||
| GenBank mRNA | AF326484.1 | - | 944 | 112 | ||
Set Membership: |
|