Assay Details
| Gene Symbol: lox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112510.1 | NP_001105980.1 | 250 | 79 | ||
| GenBank mRNA | DQ335768.1 | - | 250 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lox10| Gene Symbol: lox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112510.1 | NP_001105980.1 | 250 | 79 | ||
| GenBank mRNA | DQ335768.1 | - | 250 | 79 | ||
Set Membership: |
|