Assay Details
Gene Symbol: LOC100285810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158700.1 | NP_001152172.1 | 854 | 111 | ||
GenBank mRNA | BT062644.1 | - | 755 | 111 | ||
EU974095.1 | - | 854 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
