Assay Details
Gene Symbol: LOC100285309 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158202.1 | NP_001151674.1 | 355 | 83 | ||
GenBank mRNA | EU971687.1 | - | 355 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
