Assay Details
| Gene Symbol: LOC100282473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155382.1 | NP_001148854.1 | 251 | 93 | ||
| GenBank mRNA | EU961029.1 | - | 251 | 93 | ||
More Information
Set Membership: |
|

LOC100282473
Genomic Map