Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| GABRA2 - gamma-aminobutyric acid type A receptor alpha2 subunit | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000807.2 | Intron | NP_000798.2 | ||||
| NM_001114175.1 | Intron | NP_001107647.1 | ||||
| NM_001286827.1 | Intron | NP_001273756.1 | ||||
| XM_005248080.3 | Intron | XP_005248137.1 | ||||
| XM_011513675.1 | Intron | XP_011511977.1 | ||||
| XM_011513676.1 | Intron | XP_011511978.1 | ||||
| XM_017007982.1 | Intron | XP_016863471.1 | ||||
| XM_017007983.1 | Intron | XP_016863472.1 | ||||

GABRA2
Genomic Map