Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| SLC1A1 - solute carrier family 1 member 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004170.5 | 353 | Intron | NP_004161.4 | |||
| XM_011518007.1 | 353 | Intron | XP_011516309.1 | |||
| XM_011518008.2 | 353 | UTR 5 | XP_011516310.1 | |||
| XM_011518009.2 | 353 | UTR 5 | XP_011516311.1 | |||
| XM_011518010.1 | 353 | Intron | XP_011516312.1 | |||
| XM_017015042.1 | 353 | Intron | XP_016870531.1 | |||
| XM_017015043.1 | 353 | Intron | XP_016870532.1 | |||

SLC1A1
Genomic Map