Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFGAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ARFGAP2 - ADP ribosylation factor GTPase activating protein 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001242832.1 | Intron | NP_001229761.1 | ||||
| NM_032389.4 | Intron | NP_115765.2 | ||||
| XM_005253166.1 | Intron | XP_005253223.1 | ||||
| XM_005253167.1 | Intron | XP_005253224.1 | ||||
| XM_005253168.2 | Intron | XP_005253225.1 | ||||
| XM_006718346.1 | Intron | XP_006718409.1 | ||||
| XM_011520405.1 | Intron | XP_011518707.1 | ||||
| XM_017018413.1 | Intron | XP_016873902.1 | ||||
| XM_017018414.1 | Intron | XP_016873903.1 | ||||
| XM_017018415.1 | Intron | XP_016873904.1 | ||||
| C11orf49 - chromosome 11 open reading frame 49 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MIR6745 - microRNA 6745 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| PACSIN3 - protein kinase C and casein kinase substrate in neurons 3 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

ARFGAP2
Genomic Map