Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf60 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| C12orf60 - chromosome 12 open reading frame 60 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_175874.3 | 3600 | Intron | NP_787070.2 | |||
| XM_005253322.4 | 3600 | Intron | XP_005253379.1 | |||
| XM_011520568.2 | 3600 | Intron | XP_011518870.1 | |||
| XM_011520569.2 | 3600 | Intron | XP_011518871.1 | |||
| XM_017018871.1 | 3600 | Intron | XP_016874360.1 | |||
| XM_017018872.1 | 3600 | Intron | XP_016874361.1 | |||
| XM_017018873.1 | 3600 | Intron | XP_016874362.1 | |||
| XM_017018874.1 | 3600 | Intron | XP_016874363.1 | |||
| H2AFJ - H2A histone family member J | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_177925.3 | 3600 | UTR 3 | NP_808760.1 | |||
| HIST4H4 - histone cluster 4, H4 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LOC105369669 - uncharacterized LOC105369669 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| WBP11 - WW domain binding protein 11 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

C12orf60
Genomic Map