Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMPK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| DMPK - dystrophia myotonica protein kinase | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| DMWD - dystrophia myotonica, WD repeat containing | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004943.1 | 1462 | Silent Mutation | GCC,GCT | A,A 472 | NP_004934.1 | |
| RSPH6A - radial spoke head 6 homolog A | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

DMPK
Genomic Map