Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42EP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| CDC42EP5 - CDC42 effector protein 5 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LENG8 - leukocyte receptor cluster member 8 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_052925.3 | Intron | NP_443157.1 | ||||
| XM_005278248.4 | Intron | XP_005278305.1 | ||||
| XM_005278249.4 | Intron | XP_005278306.1 | ||||
| XM_005278250.4 | Intron | XP_005278307.1 | ||||
| XM_005278252.4 | Intron | XP_005278309.1 | ||||
| XM_006722996.3 | Intron | XP_006723059.1 | ||||
| XM_006722997.3 | Intron | XP_006723060.1 | ||||
| XM_011526413.2 | Intron | XP_011524715.1 | ||||
| XM_011526414.2 | Intron | XP_011524716.1 | ||||
| XM_011526415.2 | Intron | XP_011524717.1 | ||||
| LENG8-AS1 - LENG8 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LENG9 - leukocyte receptor cluster member 9 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

CDC42EP5
Genomic Map