Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| LSS - lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MCM3AP - minichromosome maintenance complex component 3 associated protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_003906.4 | Intron | NP_003897.2 | ||||
| XM_005261203.4 | Intron | XP_005261260.1 | ||||
| XM_005261204.4 | Intron | XP_005261261.1 | ||||
| XM_005261205.3 | Intron | XP_005261262.1 | ||||
| MCM3AP-AS1 - MCM3AP antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

LSS
Genomic Map