Assay Details
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
MCM3AP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| MCM3AP - minichromosome maintenance complex component 3 associated protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_003906.4 | 2085 | Missense Mutation | TGG,TTG | W,L 502 | NP_003897.2 | |
| XM_005261203.4 | 2085 | Missense Mutation | TGG,TTG | W,L 502 | XP_005261260.1 | |
| XM_005261204.4 | 2085 | Missense Mutation | TGG,TTG | W,L 502 | XP_005261261.1 | |
| XM_005261205.3 | 2085 | Missense Mutation | TGG,TTG | W,L 502 | XP_005261262.1 | |
| YBEY - ybeY metallopeptidase (putative) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

MCM3AP
Genomic Map