Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
C21orf58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| C21orf58 - chromosome 21 open reading frame 58 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001286462.1 | Intron | NP_001273391.1 | ||||
| NM_001286463.1 | Intron | NP_001273392.1 | ||||
| NM_001286476.1 | Intron | NP_001273405.1 | ||||
| NM_001286477.1 | Intron | NP_001273406.1 | ||||
| NM_058180.4 | Intron | NP_478060.2 | ||||
| XM_005261148.4 | Intron | XP_005261205.1 | ||||
| XM_006724018.3 | Intron | XP_006724081.1 | ||||
| XM_011529619.2 | Intron | XP_011527921.1 | ||||
| XM_011529620.2 | Intron | XP_011527922.1 | ||||
| XM_011529621.2 | Intron | XP_011527923.1 | ||||
| XM_011529623.2 | Intron | XP_011527925.1 | ||||
| XM_011529624.2 | Intron | XP_011527926.1 | ||||
| XM_011529625.2 | Intron | XP_011527927.1 | ||||
| XM_011529626.2 | Intron | XP_011527928.1 | ||||
| XM_011529627.2 | Intron | XP_011527929.1 | ||||
| XM_011529628.1 | Intron | XP_011527930.1 | ||||
| XM_017028388.1 | Intron | XP_016883877.1 | ||||
| XM_017028389.1 | Intron | XP_016883878.1 | ||||
| XM_017028390.1 | Intron | XP_016883879.1 | ||||
| XM_017028391.1 | Intron | XP_016883880.1 | ||||
| XM_017028392.1 | Intron | XP_016883881.1 | ||||
| XM_017028393.1 | Intron | XP_016883882.1 | ||||
| PCNT - pericentrin | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| YBEY - ybeY metallopeptidase (putative) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

C21orf58
Genomic Map