Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
C21orf58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| PCNT - pericentrin | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001315529.1 | 229 | Intron | NP_001302458.1 | |||
| NM_006031.5 | 229 | Intron | NP_006022.3 | |||
| XM_005261124.4 | 229 | Intron | XP_005261181.1 | |||
| XM_011529594.2 | 229 | Intron | XP_011527896.1 | |||
| XM_017028362.1 | 229 | Intron | XP_016883851.1 | |||
| XM_017028363.1 | 229 | Intron | XP_016883852.1 | |||
| YBEY - ybeY metallopeptidase (putative) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

C21orf58
Genomic Map