Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| C4orf47 - chromosome 4 open reading frame 47 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001114357.1 | Intron | NP_001107829.1 | ||||
| XM_005263019.3 | Intron | XP_005263076.1 | ||||
| XM_011531989.2 | Intron | XP_011530291.1 | ||||
| XM_017008236.1 | Intron | XP_016863725.1 | ||||
| XM_017008237.1 | Intron | XP_016863726.1 | ||||
| XM_017008238.1 | Intron | XP_016863727.1 | ||||
| XM_017008239.1 | Intron | XP_016863728.1 | ||||
| XM_017008240.1 | Intron | XP_016863729.1 | ||||
| XM_017008241.1 | Intron | XP_016863730.1 | ||||
| XM_017008242.1 | Intron | XP_016863731.1 | ||||
| CCDC110 - coiled-coil domain containing 110 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001145411.1 | Intron | NP_001138883.1 | ||||
| NM_152775.3 | Intron | NP_689988.1 | ||||
| XM_005262891.3 | Intron | XP_005262948.1 | ||||
| XM_006714172.2 | Intron | XP_006714235.1 | ||||
| XM_011531827.2 | Intron | XP_011530129.1 | ||||
| XM_011531828.2 | Intron | XP_011530130.1 | ||||
| XM_011531829.2 | Intron | XP_011530131.1 | ||||
| XM_017007987.1 | Intron | XP_016863476.1 | ||||
| XM_017007988.1 | Intron | XP_016863477.1 | ||||

C4orf47
Genomic Map