Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| C4orf47 - chromosome 4 open reading frame 47 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| CCDC110 - coiled-coil domain containing 110 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001145411.1 | Intron | NP_001138883.1 | ||||
| NM_152775.3 | Intron | NP_689988.1 | ||||
| XM_005262891.3 | Intron | XP_005262948.1 | ||||
| XM_006714172.2 | Intron | XP_006714235.1 | ||||
| XM_011531827.2 | Intron | XP_011530129.1 | ||||
| XM_011531828.2 | Intron | XP_011530130.1 | ||||
| XM_011531829.2 | Intron | XP_011530131.1 | ||||
| XM_017007987.1 | Intron | XP_016863476.1 | ||||
| XM_017007988.1 | Intron | XP_016863477.1 | ||||

C4orf47
Genomic Map