Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMKMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| CAMKMT - calmodulin-lysine N-methyltransferase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_024766.4 | Intron | NP_079042.1 | ||||
| XM_011533111.2 | Intron | XP_011531413.1 | ||||
| XM_011533113.2 | Intron | XP_011531415.1 | ||||
| XM_017004970.1 | Intron | XP_016860459.1 | ||||
| XM_017004971.1 | Intron | XP_016860460.1 | ||||
| XM_017004972.1 | Intron | XP_016860461.1 | ||||
| XM_017004973.1 | Intron | XP_016860462.1 | ||||
| XM_017004974.1 | Intron | XP_016860463.1 | ||||
| XM_017004975.1 | Intron | XP_016860464.1 | ||||
| XM_017004976.1 | Intron | XP_016860465.1 | ||||
| XM_017004977.1 | Intron | XP_016860466.1 | ||||
| XM_017004978.1 | Intron | XP_016860467.1 | ||||
| XM_017004979.1 | Intron | XP_016860468.1 | ||||
| XM_017004980.1 | Intron | XP_016860469.1 | ||||
| XM_017004981.1 | Intron | XP_016860470.1 | ||||
| XM_017004982.1 | Intron | XP_016860471.1 | ||||
| XM_017004983.1 | Intron | XP_016860472.1 | ||||
| PREPL - prolyl endopeptidase-like | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

CAMKMT
Genomic Map