Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
STARD13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| STARD13 - StAR related lipid transfer domain containing 13 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001243466.1 | Intron | NP_001230395.1 | ||||
| NM_001243474.1 | Intron | NP_001230403.1 | ||||
| NM_001243476.2 | Intron | NP_001230405.1 | ||||
| NM_052851.2 | Intron | NP_443083.1 | ||||
| NM_178006.3 | Intron | NP_821074.1 | ||||
| NM_178007.2 | Intron | NP_821075.1 | ||||
| XM_011535299.2 | Intron | XP_011533601.1 | ||||
| XM_017020834.1 | Intron | XP_016876323.1 | ||||
| XM_017020835.1 | Intron | XP_016876324.1 | ||||
| STARD13-AS - STARD13 antisense RNA | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

STARD13
Genomic Map