Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBBP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| RBBP4 - RB binding protein 4, chromatin remodeling factor | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| ZBTB8OS - zinc finger and BTB domain containing 8 opposite strand | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001308135.1 | Intron | NP_001295064.1 | ||||
| NM_001308136.1 | Intron | NP_001295065.1 | ||||
| NM_001308137.1 | Intron | NP_001295066.1 | ||||
| NM_001308138.1 | Intron | NP_001295067.1 | ||||
| NM_001308139.1 | Intron | NP_001295068.1 | ||||
| NM_001308140.1 | Intron | NP_001295069.1 | ||||
| NM_001308141.1 | Intron | NP_001295070.1 | ||||
| NM_178547.3 | Intron | NP_848642.1 | ||||
| XM_011541327.2 | Intron | XP_011539629.1 | ||||
| XM_011541328.1 | Intron | XP_011539630.1 | ||||
| XM_011541329.2 | Intron | XP_011539631.1 | ||||
| XM_011541330.2 | Intron | XP_011539632.1 | ||||
| XM_011541334.2 | Intron | XP_011539636.1 | ||||
| XM_011541335.2 | Intron | XP_011539637.1 | ||||
| XM_011541337.2 | Intron | XP_011539639.1 | ||||
| XM_011541338.2 | Intron | XP_011539640.1 | ||||
| XM_017001134.1 | Intron | XP_016856623.1 | ||||
| XM_017001135.1 | Intron | XP_016856624.1 | ||||
| XM_017001136.1 | Intron | XP_016856625.1 | ||||
| XM_017001137.1 | Intron | XP_016856626.1 | ||||
| XM_017001138.1 | Intron | XP_016856627.1 | ||||

RBBP4
Genomic Map